Dibutylbis(2,4-pentandionato-O,O')zinn, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

520586-500ML
SIAL520586-500MLEA 383 EUR
SIAL520586-500ML
Dibutylbis(2,4-pentandionato-O,O')zinn, Sigma-Aldrich®
Dibutylbis(2,4-pentandionato-O,O')zinn
Formel: C₁₈H₃₂O₄Sn
MDL: MFCD00077994
CAS-Nummer: 22673-19-4
EINECS: 245-152-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR