Platin(II)chlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Platinum dichloride
M-Clarity™ Qualitätsstufe = MQ100
520632-1G
520632-5G
SIAL520632-1GEA
465
EUR
SIAL520632-1G
SIAL520632-5G
Platin(II)chlorid, Sigma-Aldrich®
Platin(II)chlorid
Formel:
Cl₂Pt Molecular Weight: 265.99 g/mol Schmelzpunkt: 581 Dichte: 6.05 g/cm³ (20 °C) |
MDL:
MFCD00011181 CAS-Nummer: 10025-65-7 EINECS: 233-034-1 UN: 3260 ADR: 8 III Merck Index: 12,07687 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...