Iridium(III)chlorid, Sigma-Aldrich®

Lieferant: Merck

520721-1G
SIAL520721-1GEA 308 EUR
SIAL520721-1G
Iridium(III)chlorid, Sigma-Aldrich®
Iridium(III)chlorid
Formel: Cl₃Ir
Molecular Weight: 298,58 g/mol
Dichte: 5,3 g/cm³ (20 °C)
MDL: MFCD00011063
CAS-Nummer: 10025-83-9
EINECS: 233-044-6
UN: 3260
ADR: 8,II
Merck Index: 12,05101

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR