2-Brombenzaldehyddiethylacetal, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Brom-α,α-diethoxytoluol, 1-Brom-2-(diethoxymethyl)benzol

M-Clarity™ Qualitätsstufe = MQ100

520942-50ML
SIAL520942-50MLEA 137 EUR
SIAL520942-50ML
2-Brombenzaldehyddiethylacetal, Sigma-Aldrich®
2-Brombenzaldehyddiethylacetal
Formel: C₁₁H₁₅BrO₂
Molecular Weight: 259,14 g/mol
MDL: MFCD01075696
CAS-Nummer: 35822-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR