1-Butyl-4-ethinylbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Ethinyl-1-butylbenzol, (4-Butylphenyl)ethin, 4-Butyl-1-ethinylbenzol, (4-Butylphenyl)acetylen, 1-Ethinyl-4-butylbenzol

M-Clarity™ Qualitätsstufe = MQ100

521086-5G
SIAL521086-5GEA 60.5 EUR
SIAL521086-5G
1-Butyl-4-ethinylbenzol, Sigma-Aldrich®
1-Butyl-4-ethinylbenzol
Formel: C₁₂H₁₄
Molecular Weight: 158,24 g/mol
MDL: MFCD00173885
CAS-Nummer: 79887-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR