2-(Methylthio)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

521248-10G
SIAL521248-10GEA 88.1 EUR
SIAL521248-10G
2-(Methylthio)phenylboronsäure, Sigma-Aldrich®
2-(Methylthio)phenylboronsäure
Formel: C₇H₉BO₂S
MDL: MFCD01318165
CAS-Nummer: 168618-42-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR