3-(Trimethylsilylethinyl)pyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-((Trimethylsilyl)ethynyl)pyridine

M-Clarity™ Qualitätsstufe = MQ100

521264-5G
SIAL521264-5GEA 55.2 EUR
SIAL521264-5G
3-(Trimethylsilylethinyl)pyridin, Sigma-Aldrich®
3-(Trimethylsilylethinyl)pyridin
Formel: C₁₀H₁₃NSi
Molecular Weight: 175,31 g/mol
MDL: MFCD01863744
CAS-Nummer: 80673-00-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR