Tantaltris(diethylamido)-tert-butylimid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

521280-5ML
SIAL521280-5MLEA 500 EUR
SIAL521280-5ML
Tantaltris(diethylamido)-tert-butylimid, Sigma-Aldrich®
Tantaltris(diethylamido)-tert-butylimid
Formel: C₁₆H₃₉N₄Ta
MDL: MFCD02684506
CAS-Nummer: 169896-41-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR