2,4-Dichlorphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4-Dichlorbenzolboronsäure, Dihydroxy(2,4-dichlorphenyl)boran, (2,4-Dichlorphenyl)metaborsäure

M-Clarity™ Qualitätsstufe = MQ100

521388-5G
SIAL521388-5GEA 41.5 EUR
SIAL521388-5G
2,4-Dichlorphenylboronsäure, Sigma-Aldrich®
2,4-Dichlorphenylboronsäure
Formel: C₆H₅BCl₂O₂
Molecular Weight: 190,82 g/mol
MDL: MFCD00013930
CAS-Nummer: 68716-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR