1-Chlor-6-iodhexan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

521582-10G
SIAL521582-10GEA 112 EUR
SIAL521582-10G
1-Chlor-6-iodhexan, Sigma-Aldrich®
1-Chlor-6-iodhexan
Formel: C₆H₁₂ClI
Molecular Weight: 246,52 g/mol
MDL: MFCD00039417
CAS-Nummer: 34683-73-3
EINECS: 252-144-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR