(R)-3-(Boc-amino)-3-(3-methoxyphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-3-((tert-Butoxycarbonyl)amino)-3-(3-methoxyphenyl)propanoic acid

52169-500MG-F
SIAL52169-500MG-FEA 350 EUR
SIAL52169-500MG-F
(R)-3-(Boc-amino)-3-(3-methoxyphenyl)propionsäure, Sigma-Aldrich®
(R)-3-(Boc-amino)-3-(3-methoxyphenyl)propionsäure
Formel: C₁₅H₂₁NO₅
Molecular Weight: 295,34 g/mol
MDL: MFCD03427937
CAS-Nummer: 500788-86-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR