6-Ethoxy-2-naphthalinboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (6-Ethoxynaphthalen-2-yl)boronic acid

M-Clarity™ Qualitätsstufe = MQ100

521906-5G
SIAL521906-5GEA 291 EUR
SIAL521906-5G
6-Ethoxy-2-naphthalinboronsäure, Sigma-Aldrich®
6-Ethoxy-2-naphthaleneboronic acid
Formel: C₁₂H₁₃BO₃
Molecular Weight: 216,04 g/mol
MDL: MFCD03427009
CAS-Nummer: 352525-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR