1,8-Dihydroxy-4,5-dinitro-9,10-anthrachinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,8-Dihydroxy-4,5-dinitroanthraquinone

M-Clarity™ Qualitätsstufe = MQ100

522546-5G
SIAL522546-5GEA 39.2 EUR
SIAL522546-5G
1,8-Dihydroxy-4,5-dinitro-9,10-anthrachinon, Sigma-Aldrich®
1,8-Dihydroxy-4,5-dinitro-9,10-anthrachinon
Formel: C₁₄H₆N₂O₈
Molecular Weight: 330,21 g/mol
MDL: MFCD00019149
CAS-Nummer: 81-55-0
EINECS: 201-360-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR