2',6'-Dichlor-3'-fluoracetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

522945-5G
SIAL522945-5GEA 64.8 EUR
SIAL522945-5G
2',6'-Dichlor-3'-fluoracetophenon, Sigma-Aldrich®
2',6'-Dichlor-3'-fluoracetophenon
Formel: C₈H₅Cl₂FO
Molecular Weight: 207,03 g/mol
MDL: MFCD02093760
CAS-Nummer: 290835-85-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR