5-Fluor-2-methoxybenzoesäure, Sigma-Aldrich®

Lieferant: Merck

523097-5G
SIAL523097-5GEA 197 EUR
SIAL523097-5G
5-Fluor-2-methoxybenzoesäure, Sigma-Aldrich®
5-Fluor-2-methoxybenzoesäure
Formel: C₈H₇FO₃
Molecular Weight: 170,14 g/mol
MDL: MFCD00671765
CAS-Nummer: 394-04-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR