5-Fluor-o-anisaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Fluor-2-anisaldehyd, 5-Fluor-2-methoxybenzaldehyd

M-Clarity™ Qualitätsstufe = MQ100

523100-1G
SIAL523100-1GEA 25 EUR
SIAL523100-1G
5-Fluor-o-anisaldehyd, Sigma-Aldrich®
5-Fluor-o-anisaldehyd
Formel: C₈H₇FO₂
Molecular Weight: 154,14 g/mol
MDL: MFCD00143458
CAS-Nummer: 19415-51-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR