2-(Methylthio)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

523119-5G
SIAL523119-5GEA 272 EUR
SIAL523119-5G
2-(Methylthio)benzaldehyd, Sigma-Aldrich®
2-(Methylthio)benzaldehyde
Formel: C₈H₈OS
Molecular Weight: 152,2135 g/mol
MDL: MFCD00196822
CAS-Nummer: 7022-45-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR