3-Chlor-2-methylanisol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chloro-6-methoxytoluene, 1-Chloro-3-methoxy-2-methylbenzene

M-Clarity™ Qualitätsstufe = MQ100

523232-10G
SIAL523232-10GEA 121 EUR
SIAL523232-10G
3-Chlor-2-methylanisol, Sigma-Aldrich®
3-Chlor-2-methylanisol
Formel: C₈H₉ClO
Molecular Weight: 156,61 g/mol
MDL: MFCD00070772
CAS-Nummer: 3260-88-6
EINECS: 221-862-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR