4-Brom-2-iodanilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

523267-5G
SIAL523267-5GEA 85.4 EUR
SIAL523267-5G
4-Brom-2-iodanilin, Sigma-Aldrich®
4-Brom-2-iodanilin
Formel: C₆H₅BrIN
Molecular Weight: 297,92 g/mol
MDL: MFCD02093764
CAS-Nummer: 66416-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR