4-[(Trimethylsilyl)ethinyl]benzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-((Trimethylsilyl)ethynyl)benzaldehyde, 4-(Trimethylsilylethynyl)benzaldehyde

523380-5G
SIAL523380-5GEA 202 EUR
SIAL523380-5G
4-[(Trimethylsilyl)ethinyl]benzaldehyd, Sigma-Aldrich®
4-[(Trimethylsilyl)ethinyl]benzaldehyd
Formel: C₁₂H₁₄OSi
Molecular Weight: 202,33 g/mol
MDL: MFCD02093765
CAS-Nummer: 77123-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR