4-Iodbenzylalkohol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-Iodophenyl)methanol

M-Clarity™ Qualitätsstufe = MQ100

523496-5G
SIAL523496-5GEA 115 EUR
SIAL523496-5G
4-Iodbenzylalkohol, Sigma-Aldrich®
4-Iodbenzylalkohol
Formel: C₇H₇IO
Molecular Weight: 234,04 g/mol
MDL: MFCD01732720
CAS-Nummer: 18282-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR