2,3-Dichlorbenzylalkohol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: (2,3-Dichlorophenyl)methanol

523526-5G
SIAL523526-5GEA 32.7 EUR
SIAL523526-5G
2,3-Dichlorbenzylalkohol, Sigma-Aldrich®
2,3-Dichlorbenzylalkohol
Formel: C₇H₆Cl₂O
Molecular Weight: 177,03 g/mol
MDL: MFCD00238581
CAS-Nummer: 38594-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR