4-(Trimethylsilyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-(Trimethylsilyl)phenyl)boronic acid

M-Clarity™ Qualitätsstufe = MQ100

523674-5G
SIAL523674-5GEA 158 EUR
SIAL523674-5G
4-(Trimethylsilyl)phenylboronsäure, Sigma-Aldrich®
4-(Trimethylsilyl)phenylboronsäure
Formel: C₉H₁₅BO₂Si
Molecular Weight: 194,1100 g/mol
MDL: MFCD00093348
CAS-Nummer: 17865-11-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR