2,5-Dichlorbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

523917-5G
SIAL523917-5GEA 54.4 EUR
SIAL523917-5G
2,5-Dichlorbenzaldehyd, Sigma-Aldrich®
2,5-Dichlorobenzaldehyde
Formel: C₇H₄Cl₂O
Molecular Weight: 175,01 g/mol
MDL: MFCD00156140
CAS-Nummer: 6361-23-5
EINECS: 228-832-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR