2,5-Dichlor-4-nitroanilin, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
524204-10G
SIAL524204-10GEA
160
EUR
SIAL524204-10G
2,5-Dichlor-4-nitroanilin, Sigma-Aldrich®
2,5-Dichlor-4-nitroanilin
Formel:
C₆H₄Cl₂N₂O₂ Molecular Weight: 207,02 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025157 CAS-Nummer: 6627-34-5 EINECS: 229-591-5 UN: 2237 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...