Methyldiphenylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Methyl-N-phenylaniline

M-Clarity™ Qualitätsstufe = MQ100

524212-25G
SIAL524212-25GEA 185 EUR
SIAL524212-25G
Methyldiphenylamin, Sigma-Aldrich®
Methyldiphenylamin
Formel: C₁₃H₁₃N
Molecular Weight: 183,25 g/mol
MDL: MFCD00041900
CAS-Nummer: 552-82-9
EINECS: 209-023-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR