5-Iodsalicylaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Hydroxy-5-iodobenzaldehyde

524328-1G
SIAL524328-1GEA 76.1 EUR
SIAL524328-1G
5-Iodsalicylaldehyd, Sigma-Aldrich®
5-Iodsalicylaldehyd
Formel: C₇H₅IO₂
Molecular Weight: 248,0200 g/mol
MDL: MFCD00723501
CAS-Nummer: 1761-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR