6-Bromindol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
6-Bromo-1H-indole
M-Clarity™ Qualitätsstufe = MQ200
524344-1G
524344-5G
SIAL524344-1GEA
81.5
EUR
SIAL524344-1G
SIAL524344-5G
6-Bromindol, Sigma-Aldrich®
6-Bromindol
Formel:
C₈H₆BrN Lagertemperatur: Raumtemperatur |
MDL:
MFCD00238550 CAS-Nummer: 52415-29-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...