cis-1,4-Hexadien, Sigma-Aldrich®

Lieferant: Merck

52435-5ML
SIAL52435-5MLEA 112 EUR
SIAL52435-5ML
cis-1,4-Hexadien, Sigma-Aldrich®
cis-1,4-Hexadiene
Formel: C₆H₁₀
CAS-Nummer: 7318-67-4
EINECS: 230-783-6
UN: 2458
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR