11-Hydroxyundecansäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

524395-1G
SIAL524395-1GEA 160 EUR
SIAL524395-1G
11-Hydroxyundecansäure, Sigma-Aldrich®
Formel: C₁₁H₂₂O₃
MDL: MFCD00041564
CAS-Nummer: 3669-80-5
EINECS: 222-932-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR