3,5-Dinitro-o-toluamid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3,5-Dinitro-2-methylbenzoesäureamid, 3,5-Dinitro-o-tolylsäureamid, Dinitolmid
M-Clarity™ Qualitätsstufe = MQ100
524417-50G
SIAL524417-50GEA
165
EUR
SIAL524417-50G
3,5-Dinitro-o-toluamid, Sigma-Aldrich®
3,5-Dinitro-o-toluamid
Formel:
C₈H₇N₃O₅ Molecular Weight: 225,16 g/mol Schmelzpunkt: 180 °C |
MDL:
MFCD00072065 CAS-Nummer: 148-01-6 EINECS: 205-706-4 Merck Index: 12,03321 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...