Tris[N,N-bis(trimethylsilyl)amid]yttrium(III), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

524514-5G
SIAL524514-5GEA 463 EUR
SIAL524514-5G
Tris[N,N-bis(trimethylsilyl)amid]yttrium(III), Sigma-Aldrich®
Tris[N,N-bis(trimethylsilyl)amid]yttrium(III)
Formel: C₁₈H₅₄N₃Si₆Y
Molecular Weight: 570,06 g/mol
MDL: MFCD00210649
CAS-Nummer: 41836-28-6
UN: 3396
ADR: 4.3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR