2,3,5-Trifluorphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

524697-5G
SIAL524697-5GEA 165 EUR
SIAL524697-5G
2,3,5-Trifluorphenylboronsäure, Sigma-Aldrich®
2,3,5-Trifluorphenylboronsäure
Formel: C₆H₄BF₃O₂
Molecular Weight: 175,9 g/mol
MDL: MFCD01863166
CAS-Nummer: 247564-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR