2-Methoxy-4-nitrobenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

524751-5G
SIAL524751-5GEA 34.5 EUR
SIAL524751-5G
2-Methoxy-4-nitrobenzonitril, Sigma-Aldrich®
2-Methoxy-4-nitrobenzonitril
Formel: C₈H₆N₂O₃
Molecular Weight: 178,15 g/mol
MDL: MFCD02093781
CAS-Nummer: 101084-96-2
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR