2,4,5-Trifluor-3-methoxybenzoesäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Moxidectin Impurity 5, 3-Methoxy-2,4,5-trifluorobenzoic acid

M-Clarity™ Qualitätsstufe = MQ100

524824-5G
SIAL524824-5GEA 32 EUR
SIAL524824-5G
2,4,5-Trifluor-3-methoxybenzoesäure, Sigma-Aldrich®
2,4,5-Trifluor-3-methoxybenzoesäure
Formel: C₈H₅F₃O₃
Molecular Weight: 206,12 g/mol
MDL: MFCD00153201
CAS-Nummer: 112811-65-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR