1-Tritylimidazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Clotrimazole Impurity F, 1-Trityl-1H-imidazole

524891-25G
SIAL524891-25GEA 477 EUR
SIAL524891-25G
1-Tritylimidazol, Sigma-Aldrich®
1-Tritylimidazol
Formel: C₂₂H₁₈N₂
Molecular Weight: 310,40 g/mol
MDL: MFCD00229427
CAS-Nummer: 15469-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR