4-Aminoindol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Indol-4-ylamin, 4-Indolamin, 4-Amino-1H-indol

525022-500MG
SIAL525022-500MGEA 129 EUR
SIAL525022-500MG
4-Aminoindol, Sigma-Aldrich®
4-Aminoindol
Formel: C₈H₈N₂
Molecular Weight: 132,16 g/mol
Schmelzpunkt: 108 °C
MDL: MFCD01076559
CAS-Nummer: 5192-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR