1H-Pyrazol-4-boronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

525057-1G 525057-5G
SIAL525057-1GEA 49.9 EUR
SIAL525057-1G SIAL525057-5G
1H-Pyrazol-4-boronsäurepinakolester, Sigma-Aldrich®
1H-Pyrazol-4-boronsäurepinakolester
Formel: C₉H₁₅BN₂O₂
MDL: MFCD03453063
CAS-Nummer: 269410-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR