4-Chlor-2-methoxybenzylalkohol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-Chloro-2-methoxyphenyl)methanol

M-Clarity™ Qualitätsstufe = MQ100

525715-5G
SIAL525715-5GEA 116 EUR
SIAL525715-5G
4-Chlor-2-methoxybenzylalkohol, Sigma-Aldrich®
4-Chlor-2-methoxybenzylalkohol
Formel: C₈H₉ClO₂
Molecular Weight: 172,61 g/mol
MDL: MFCD02683547
CAS-Nummer: 55685-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR