trans-2-Aminocyclopentanol Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

525863-5G
SIAL525863-5GEA 234 EUR
SIAL525863-5G
trans-2-Aminocyclopentanol Hydrochlorid, Sigma-Aldrich®
trans-2-Aminocyclopentanol Hydrochlorid
Formel: C₅H₁₂ClNO
Molecular Weight: 137,607 g/mol
MDL: MFCD02683551
CAS-Nummer: 31775-67-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR