trans-2-(4-Biphenyl)vinylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (E)-(2-([1,1'-Biphenyl]-4-yl)vinyl)boronic acid

M-Clarity™ Qualitätsstufe = MQ100

526045-1G
SIAL526045-1GEA 107 EUR
SIAL526045-1G
trans-2-(4-Biphenyl)vinylboronsäure, Sigma-Aldrich®
trans-2-(4-Biphenyl)vinylboronsäure
Formel: C₁₄H₁₃BO₂
Molecular Weight: 224,600 g/mol
MDL: MFCD04974059
CAS-Nummer: 352530-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR