Diethyl(4-chlorbenzyl)phosphonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diethyl 4-chlorobenzylphosphonate

M-Clarity™ Qualitätsstufe = MQ100

526061-10G
SIAL526061-10GEA 73.7 EUR
SIAL526061-10G
Diethyl(4-chlorbenzyl)phosphonat, Sigma-Aldrich®
Diethyl(4-chlorbenzyl)phosphonat
Formel: C₁₁H₁₆ClO₃P
Molecular Weight: 262,67 g/mol
MDL: MFCD00051568
CAS-Nummer: 39225-17-7
EINECS: 254-365-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR