5-Fluorsalicylaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Fluoro-2-hydroxybenzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

526274-1G
SIAL526274-1GEA 67.2 EUR
SIAL526274-1G
5-Fluorsalicylaldehyd, Sigma-Aldrich®
5-Fluorsalicylaldehyd
Formel: C₇H₅FO₂
Molecular Weight: 140,11 g/mol
MDL: MFCD01090997
CAS-Nummer: 347-54-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR