2-Chlor-3,6-difluorbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

526428-5G
SIAL526428-5GEA 99.6 EUR
SIAL526428-5G
2-Chlor-3,6-difluorbenzaldehyd, Sigma-Aldrich®
2-Chloro-3,6-difluorobenzaldehyde
Formel: C₇H₃ClF₂O
Molecular Weight: 176,55 g/mol
MDL: MFCD01631443
CAS-Nummer: 261762-39-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR