2,4,6-Trichlorbenzylalkohol, Sigma-Aldrich®

Lieferant: Merck

526436-5G
SIAL526436-5GEA 480 EUR
SIAL526436-5G
2,4,6-Trichlorbenzylalkohol, Sigma-Aldrich®
2,4,6-Trichlorbenzylalkohol
Formel: C₇H₅Cl₃O
Molecular Weight: 211,47 g/mol
MDL: MFCD03093891
CAS-Nummer: 217479-60-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR