2,4,5-Trimethylbenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

526444-5G
SIAL526444-5GEA 87.7 EUR
SIAL526444-5G
2,4,5-Trimethylbenzoesäure, Sigma-Aldrich®
2,4,5-Trimethylbenzoic acid
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,2 g/mol
MDL: MFCD00075863
CAS-Nummer: 528-90-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR