2,4,5-Trimethylbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

526452-1G
SIAL526452-1GEA 53 EUR
SIAL526452-1G
2,4,5-Trimethylbenzaldehyd, Sigma-Aldrich®
2,4,5-Trimethylbenzaldehyde
Formel: C₁₀H₁₂O
MDL: MFCD00017713
CAS-Nummer: 5779-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR