4-Methyl-1-naphthoesäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Methyl-1-naphthalenecarboxylic acid

526533-10G
SIAL526533-10GEA 111 EUR
SIAL526533-10G
4-Methyl-1-naphthoesäure, Sigma-Aldrich®
4-Methyl-1-naphthoesäure
Formel: C₁₂H₁₀O₂
MDL: MFCD00671557
CAS-Nummer: 4488-40-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR