2,4,5-Trifluorbenzylalkohol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

526541-5G
SIAL526541-5GEA 22.7 EUR
SIAL526541-5G
2,4,5-Trifluorbenzylalkohol, Sigma-Aldrich®
2,4,5-Trifluorbenzylalkohol
Formel: C₇H₅F₃O
Molecular Weight: 162,11 g/mol
MDL: MFCD00061206
CAS-Nummer: 144284-25-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR