3-Chlor-2,6-difluorbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

526592-1G
SIAL526592-1GEA 40.2 EUR
SIAL526592-1G
3-Chlor-2,6-difluorbenzaldehyd, Sigma-Aldrich®
3-Chloro-2,6-difluorobenzaldehyde
Formel: C₇H₃ClF₂O
Molecular Weight: 176,55 g/mol
MDL: MFCD01631323
CAS-Nummer: 190011-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR